Learn More
Medchemexpress LLC CD137/4-1BB protein, rhesus macaque (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000293945
Encompass_Preferred
Recombinant extracellular domain of rhesus macaque CD137/4-1BB expressed in HEK293 cells with a C-terminal His tag. Lyophilized, high-purity protein formulated in PBS with low endotoxin, intended for research applications in immunology, receptor-ligand assays, and functional studies. Supplied with documentation including datasheet, COA, and SDS.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg.
  • Lyophilized powder formulated from 0.22 μm filtered PBS, pH 7.4.
  • Produced in mammalian cells for proper folding and glycosylation.
  • C-terminal His tag for purification and detection.
  • Stable when stored frozen; aliquot and add carrier protein for long-term storage.
Catalog No. 50-002-93945 Supplier Medchemexpress LLC Supplier No. HYP7619910UG
Add to Cart
Add to Cart
Recombinant extracellular domain of rhesus macaque CD137/4-1BB expressed in HEK293 cells with a C-terminal His tag. Lyophilized, high-purity protein formulated in PBS with low endotoxin, intended for research applications in immunology, receptor-ligand assays, and functional studies. Supplied with documentation including datasheet, COA, and SDS.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg.
  • Lyophilized powder formulated from 0.22 μm filtered PBS, pH 7.4.
  • Produced in mammalian cells for proper folding and glycosylation.
  • C-terminal His tag for purification and detection.
  • Stable when stored frozen; aliquot and add carrier protein for long-term storage.