Learn More
Enzo Life Sciences CD137 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522031C050
Encompass_Preferred
Alternative name: 4-1BB, TNFRSF 9. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The cysteine-rich region of human CD137 (4-1BB) (aa 1-186) is fused to the Fc portion of human IgG1. Specificity: Binds human CD137L (4-1BBL). Long Term Storage: -20°C. Applications: ELISA, FUNC .
Catalog No. 50-200-9379