Learn More
Biotium CD137L / 4-1BBL / TNFSF9(CD137L/1547), CF647 conjugate, 0.1mg/mL
Supplier:  Biotium BNC471547100
Encompass_Preferred
TNFSF9 / 4-1BBL is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This transmembrane cytokine is a bidirectional signal transducer that acts as a ligand for TNFRSF9 / 4-1BB, which is a costimulatory receptor molecule in T lymphocytes. This cytokine and its receptor are involved in the antigen presentation process and in the generation of cytotoxic T cells. The receptor TNFRSF9 / 4-1BB is absent from resting T lymphocytes but rapidly expressed upon antigenic stimulation. It has been shown to reactivate anergic T lymphocytes in addition to promoting T lymphocyte proliferation. This cytokine is required for the optimal CD8 responses in CD8 T cells. This cytokine is expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interaction. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluo
Catalog No. 50-252-4819