Learn More
AdipoGen anti-CD137L (human), mAb (41B436) (50 µg)
SDP
Supplier:  AdipoGen AG20A0031C050
Encompass_Preferred
Monoclonal Antibody. Recognizes extracellular domain of human CD137L. Does not cross-react with mouse CD137L. Isotype: Mouse IgG1kappa. Clone: 41B436. Applications: ELISA, FACS, ICC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The CD137 ligand (CD137L, 4-1BBL) is a member of the tumor necrosis factor (TNF) family. CD137L is a type II, transmembrane protein found on activated macrophages, dendritic cells and mature B cells. The interaction with its receptor 4-1BB induces recruitment of TNF receptor-associated factor 1 (TRAF1) and TRAF2 and interaction with the kinase p56lck. CD137 and CD137L have been reported to be involved in tumor rejection, apoptosis, anti-viral immunity, diabetes, in T and B cell co-stimulation and modulation of the immune response.
Catalog No. 50-205-2813