Learn More
Enzo Life Sciences CD137L (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX5221113010
Encompass_Preferred
Biologically active CD137L (4-1BBL; TNFSF 9) comprising the whole extracellular tail domain (aa 49-93), which is important for the cross-linking of the protein that is essential for its biological activity. Biologically active CD137L (4-1BBL; TNFSF 9) comprising the whole extracellular tail domain (aa 49-93), which is important for the cross-linking of the protein that is essential for its biological activity. Alternative name: 4-1BBL, TNFSF 9. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS and 5% trehalose. Source: Produced in HEK 293 cells. The extracellular domain of human CD137L (aa 49-254) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG-tag. Specificity: Binds to human CD137 (4-1BB). Long Term Storage: -20°C.
Catalog No. 50-200-9430