Learn More
CAPRICO BIOTECHNOLOGIES INC CD14 Antigen Presenting Cells, APC
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4074045
Encompass_Preferred
CD14 Antigen Presenting Cells (APC) from is a monoclonal antibody that specifically binds to human CD14. CD14 is a 55 kDa GPI-anchored glycoprotein expressed on monocytes, macrophages and neutrophils. This antibody is conjugated to APC and is recommended for flow cytometry analysis of human CD14 expression on antigen presenting cells. It has been tested and validated for flow cytometry applications. The antibody is supplied as 100 tests in PBS with 0.1% BSA and 0.09% sodium azide.
Catalog No. 50-227-9023