Learn More
Enzo Life Sciences CD14 (human) monoclonal antibody (biG 2/RoMo-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804498C100
Encompass_Preferred
Clone: biG 2/RoMo-1. Isotype: IgG2a. Immunogen: Human monocytes. Species reactivity: Human. Specificity: Recognized epitope: aa 147-152 of human CD14. Applications: ELISA, Flow Cytometry, IP. Purity: Protein G-affinity purified. Formulation: Lyophilized from PBS. Contains no preservatives.
Catalog No. 50-200-9926