Learn More
Genscript Corporation CD14 His Human
Catalog No. 5000058971
Encompass
Human monocyte differentiation antigen CD14 is a pattern recognition receptor (PRR) that enhances innate immune responses CD14 was first identified as a marker of monocytes to signal intracellular responses upon bacterial encounters Given the absence of an intracellular tail CD14 was doubted to have the signaling capacities
Catalog No. 50-000-58971 Supplier Genscript Corporation Supplier No. Z04155500
Human monocyte differentiation antigen CD14 is a pattern recognition receptor (PRR) that enhances innate immune responses CD14 was first identified as a marker of monocytes to signal intracellular responses upon bacterial encounters Given the absence of an intracellular tail CD14 was doubted to have the signaling capacities