Learn More
Genscript Corporation CD14 His Mouse
Catalog No. 5000056926
Encompass
CD14 is a glycosylphosphatidylinositol (GPI)-anchored receptor known to serve as a co-receptor for several Toll-like Receptors (TLRs) both at the cell surface and in the endosomal compartment CD14 can be expressed by cells of both hematopoietic and non-hematopoietic origin as a cell membrane or secreted protein
Catalog No. 50-000-56926 Supplier Genscript Corporation Supplier No. Z04294500
CD14 is a glycosylphosphatidylinositol (GPI)-anchored receptor known to serve as a co-receptor for several Toll-like Receptors (TLRs) both at the cell surface and in the endosomal compartment CD14 can be expressed by cells of both hematopoietic and non-hematopoietic origin as a cell membrane or secreted protein