Learn More
CAPRICO BIOTECHNOLOGIES INC CD14 iFluor 700 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4034195
Encompass_Preferred
The Caprico CD14 iFluor 700 Antibody is a mouse monoclonal antibody that specifically binds to human CD14. It is conjugated to iFluor 700 and is suitable for flow cytometry, immunofluorescence microscopy and immunohistochemistry. This antibody recognizes membrane-bound and soluble CD14. CD14 is expressed primarily on monocytes/macrophages and serves as a receptor for lipopolysaccharide binding protein complexes. This antibody can be a useful tool for studying monocyte maturation and activation.
Catalog No. 50-227-9019