Learn More
Enzo Life Sciences CD14 monoclonal antibody (biG 10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804496C100
Encompass_Preferred
Clone: biG 10. Isotype: IgG1. Immunogen: Monocytes and immunoaffinity purified soluble human CD14. Species reactivity: Human Porcine, Horse, Dog, Bovine. Specificity: Recognized epitopes in human: aa 9-13 and aa 39-44. Applications: ELISA, Flow Cytometry, WB, FUNC. Purity: Protein G affinity-purified. Formulation: Lyophilized from PBS. Contains no preservatives.
Catalog No. 50-200-9924