Learn More
Enzo Life Sciences CD14 (mouse) monoclonal antibody (biG 53) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804499C100
Encompass_Preferred
Clone: biG 53. Isotype: IgG1. Immunogen: Recombinant mouse CD14. Species reactivity: Mouse. Specificity: Binds to LPS-activated mouse monocytes. Applications: ELISA, Flow Cytometry, FUNC. Purity: Protein G-affinity purified. Formulation: Lyophilized. Contains no preservatives.
Catalog No. 50-200-9927