Learn More
Enzo Life Sciences CD14 (mouse), (recombinant) (His-tag) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX201134C010
Encompass_Preferred
Concentration: 0.25mg/ml after reconstitution. Endotoxin: <0.00015EU/µg protein (LAL test; Charles River). Purity: ≥90-95% (SDS-PAGE). MW: ~45kDa. Formulation: Lyophilized from PBS, pH 7.2. Source: Produced in CHO cells. Long Term Storage: -80°C. Use/Stability: The reconstituted protein should be used immediately or stored in undiluted working aliquots at -20°C. For long term storage it is recommended to add a carrier protein (0.1% BSA). Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7931