Learn More
Beckman Coulter CD14-PB, RMO52, 0.5 mL, ASR, 0.5 mL
Catalog No. NC2954605 Shop All Beckman Coulter Products
Encompass
The CD14 antigen is a glycosyl-phosphatidylinositol-linked single-chain surface membrane glycoprotein with a molecular weight of 53-55 kDa. It functions as a high affinity receptor for the complex of LipoPolySaccharide (LPS) and the LPS-Binding Protein (LBP). It is found on cells of myelomonocytic lineage. CD14 antigen is strongly expressed on monocytes, macrophages, and weakly on neutrophils. It is also weakly expressed on B lymphocytes, but is absent from T lymphocytes, NK cells, red blood cells and platelets. It is found on Langerhans cells, follicular dendritic cells and histiocytes. *Alexa Fluor and Pacific Blue are registered trademarks of Molecular Probes, Inc.
This item is not returnable. View return policy
Catalog No. NC2954605 Supplier Beckman Coulter Supplier No. B00846
The CD14 antigen is a glycosyl-phosphatidylinositol-linked single-chain surface membrane glycoprotein with a molecular weight of 53-55 kDa. It functions as a high affinity receptor for the complex of LipoPolySaccharide (LPS) and the LPS-Binding Protein (LBP). It is found on cells of myelomonocytic lineage. CD14 antigen is strongly expressed on monocytes, macrophages, and weakly on neutrophils. It is also weakly expressed on B lymphocytes, but is absent from T lymphocytes, NK cells, red blood cells and platelets. It is found on Langerhans cells, follicular dendritic cells and histiocytes. *Alexa Fluor and Pacific Blue are registered trademarks of Molecular Probes, Inc.