Learn More
Beckman Coulter Beckman Coulter CD14-PE Conjugated Antibody, Clone RMO52, 2 mL (IM0650U)
Supplier:  Beckman Coulter IM0650U
Encompass
Beckman Coulter CD14-PE is a phycoerythrin-conjugated monoclonal antibody (clone RMO52) that specifically recognizes human CD14 for use in flow cytometry. Supplied as a 2 mL liquid reagent and classified as an Analyte Specific Reagent (ASR), it delivers bright PE fluorescence for reliable immunophenotyping.
  • Targets human CD14 expressed on monocytes and macrophages.
  • Phycoerythrin conjugate provides bright fluorescence for flow cytometry detection.
  • Supplied as a 2 mL liquid format for immediate use.
  • Mouse IgG2a isotype suitable for standard secondary controls.
  • High specificity reduces background staining in leukocyte panels.
  • Classified as an ASR for laboratory-developed tests and research use.
Catalog No. NC2411049