Learn More
Enzo Life Sciences CD14, soluble (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850302KI01
Encompass_Preferred
The CD14, soluble (human) ELISA kit is a complete assay for the quantitative determination of soluble human CD14 concentration. This kit is to be used with serum, plasma and cell culture medium from human origin. Assay Time: 2 hours + 13+/- 1 minute. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0154