Learn More
ABclonal Technology CD141/Thrombomodulin Rabbit
SDP
Catalog No. 5000006686
Encompass_Preferred
The protein encoded by this intronless gene is an endothelial-specific type I membrane receptor that binds thrombin This binding results in the activation of protein C which degrades clotting factors Va and VIIIa and reduces the amount of thrombin generated Mutations in this gene are a cause of thromboembolic disease also known as inherited thrombophilia
Catalog No. 50-000-06686 Supplier ABclonal Technology Supplier No. A22989PM20UL
The protein encoded by this intronless gene is an endothelial-specific type I membrane receptor that binds thrombin This binding results in the activation of protein C which degrades clotting factors Va and VIIIa and reduces the amount of thrombin generated Mutations in this gene are a cause of thromboembolic disease also known as inherited thrombophilia