Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 682MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Bioactive, Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 8D6. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HSB2 cells. Renal Cell, Ovarian carcinoma or Melanoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human T cell line Molt 13. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 35kDa (non-reduced); 40kDa (reduced).Chromosome: 19p13.3. Gene Symbol: BSG. Gene ID: 682. Ensemble: ENSG00000172270. Uniprot: NP_940993.
Catalog No. 50-188-5194