Learn More
ACROBiosystems Recombinant Protein;Human CD14 Protein, Fc Tag;HEK293;100UG;CD4,CD14
Supplier:  ACROBiosystems CD4H5252100UG
Encompass_Preferred
Recombinant Protein;Human CD14 Protein, Fc Tag;100UG;Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession # P08571-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7483