Learn More
ACROBiosystems Recombinant Protein;Human CD14 Protein;HEK293;100UG;CD4,CD14
Supplier:  ACROBiosystems CD4H5228100UG
Encompass_Preferred
Recombinant Protein;Human CD14 Protein;100UG;Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Met 344 (Accession # AAH10507).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.6 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8965