Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2526MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: FUT4/815. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: U937 cells, Reed-Sternberg's cells in Hodgkin's lymphoma. Cellular Staining: Cell surface, granular paranuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified human neutrophils. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~220kDa (Glycoprotein).Chromosome: 11q21. Gene Symbol: FUT4. Gene ID: 2526. Ensemble: ENSG00000196371. Uniprot: NP_002024.
Catalog No. 50-188-4725