Learn More
Biogenex Laboratories CD15 : BRA4F1 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3025M
Encompass_Preferred
CD15 (BRA4F1) reacts with human CD15 antigen present on myeloidcells, mainly granulocytes but not on B cells, T cells, monocytes,erythrocytes or platelets. It also reacts with Hodgkin's and ReedSternberg cells in individuals with Hodgkin's disease
Catalog No. 50-197-8832