Learn More
Cell Signaling Technology CD15/SSEA1 Mouse Monoclonal Antibody, Unconjugated, Human, Mouse, Monkey, 100 μL
SDP
Catalog No. 502733131
Encompass
This monoclonal antibody is designed for the detection of endogenous levels of CD15/SSEA1 protein. It is produced by immunizing BALB/c mice with the U-937 histiocytic cell line and is supplied as a liquid tissue culture supernatant. The antibody targets a lactoseries oligosaccharide antigen expressed on mouse embryonal carcinoma and embryonic stem cells, as well as early mouse embryos and human germ cells.
  • Validated for use in immunohistochemistry (paraffin), immunofluorescence (immunocytochemistry), and immunohistochemical applications.
  • Recognizes endogenous protein levels.
  • Reactive in human, mouse, and monkey samples.
  • Supplied in a formulation containing sodium azide as a preservative.
  • Stable for 6 months when stored at 4°C.
Catalog No. 50-273-3131 Supplier Cell Signaling Technology Supplier No. 54192S
This monoclonal antibody is designed for the detection of endogenous levels of CD15/SSEA1 protein. It is produced by immunizing BALB/c mice with the U-937 histiocytic cell line and is supplied as a liquid tissue culture supernatant. The antibody targets a lactoseries oligosaccharide antigen expressed on mouse embryonal carcinoma and embryonic stem cells, as well as early mouse embryos and human germ cells.
  • Validated for use in immunohistochemistry (paraffin), immunofluorescence (immunocytochemistry), and immunohistochemical applications.
  • Recognizes endogenous protein levels.
  • Reactive in human, mouse, and monkey samples.
  • Supplied in a formulation containing sodium azide as a preservative.
  • Stable for 6 months when stored at 4°C.