Learn More
AdipoGen CD152 hrecHis
SDP
Catalog No. 502562833
Encompass_Preferred
Recombinant Protein. Human CD152 aa 36-161 is fused at the C-terminus to a His-tag. Source W2. Lyophilized. Contains PBS. CD152 Cytotoxic T-lymphocyte-associated protein 4 CTLA-4 is an inhibitory receptor belonging to the CD28 immunoglobulin subfamily, expressed primarily by T-cells. Its ligands, CD80 and CD86, are typically found on the surface of antigen-presenting cells and can either bind CD28 or CTLA-4, resulting in a costimulatory or a co-inhibitory response, respectively. Because of its dampening effect, CTLA-4 is a crucial regulator of T-cell homeostasis and self-tolerance. While CD28 promotes T-cell activation and proliferation, CTLA-4 is reported to dampen T-cell responses through a variety of mechanisms. Prior to activation, conventional T-cells Tconv express low levels of CTLA-4, predominantly in intracellular compartments. Upon activation, CTLA-4 expression is upregulated and becomes increasingly detectable on the cell surface.
Catalog No. 50-256-2833 Supplier AdipoGen Supplier No. AG40B0228C050
Recombinant Protein. Human CD152 aa 36-161 is fused at the C-terminus to a His-tag. Source W2. Lyophilized. Contains PBS. CD152 Cytotoxic T-lymphocyte-associated protein 4 CTLA-4 is an inhibitory receptor belonging to the CD28 immunoglobulin subfamily, expressed primarily by T-cells. Its ligands, CD80 and CD86, are typically found on the surface of antigen-presenting cells and can either bind CD28 or CTLA-4, resulting in a costimulatory or a co-inhibitory response, respectively. Because of its dampening effect, CTLA-4 is a crucial regulator of T-cell homeostasis and self-tolerance. While CD28 promotes T-cell activation and proliferation, CTLA-4 is reported to dampen T-cell responses through a variety of mechanisms. Prior to activation, conventional T-cells Tconv express low levels of CTLA-4, predominantly in intracellular compartments. Upon activation, CTLA-4 expression is upregulated and becomes increasingly detectable on the cell surface.