Learn More
Medchemexpress LLC Cd1530 | 107430-66-0 | 98.0% | 398.49 | C27H26O3 | 5 MG
SDP
Catalog No. 5000263349
Encompass_Preferred
CD1530 is a selective retinoic acid receptor γ (RARγ) agonist supplied for research use. It is provided as a high-purity solid (C27H26O3, MW 398.49) suitable for biochemical, cellular, and in vivo studies investigating RARγ signaling, heterotopic ossification, tendon healing, and muscle pathology.
  • Selective RARγ agonist activity for targeted receptor modulation.
  • High purity solid suitable for biochemical and pharmacological studies.
  • Available in small milligram-scale packages for research applications.
  • Also offered as a DMSO solution for cell-based assays.
  • Recommended cold storage to preserve stability and activity.
Catalog No. 50-002-63349 Supplier Medchemexpress LLC Supplier No. HY1085275MG
CD1530 is a selective retinoic acid receptor γ (RARγ) agonist supplied for research use. It is provided as a high-purity solid (C27H26O3, MW 398.49) suitable for biochemical, cellular, and in vivo studies investigating RARγ signaling, heterotopic ossification, tendon healing, and muscle pathology.
  • Selective RARγ agonist activity for targeted receptor modulation.
  • High purity solid suitable for biochemical and pharmacological studies.
  • Available in small milligram-scale packages for research applications.
  • Also offered as a DMSO solution for cell-based assays.
  • Recommended cold storage to preserve stability and activity.