Learn More
ACROBiosystems ACROBiosystems Human CD155 / PVR Protein, Mouse IgG2a Fc Tag, low endotoxin
Catalog No. 501957381
Encompass_Preferred
Recombinant Protein;1MG;Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. As a result of glycosylation, the protein migrates as 70-100 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 0.1-4 ng/mL (QC tested).;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin
Catalog No. 50-195-7381 Supplier ACROBiosystems Supplier No. CD5H52541MG
Recombinant Protein;1MG;Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. As a result of glycosylation, the protein migrates as 70-100 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 0.1-4 ng/mL (QC tested).;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin