Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse CD155 / PVR Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;CD5,CD155,PVR,FLJ25946,PVS,TAGE4,HVED,NECL5,NECL-5,TAGE-4
Catalog No. 501658103
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse CD155 / PVR Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Mouse CD155, Fc Tag, Avi Tag (CD5-M82F7) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 63.5 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse TIGIT, Fc Tag (Cat. No. TIT-M5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse CD155, Fc Tag, Avi Tag (Cat. No. CD5-M82F7) with a linear range of 20-313 ng/mL (QC tested).
Catalog No. 50-165-8103 Supplier ACROBiosystems Supplier No. CD5M82F7200UG
Recombinant Protein;Biotinylated Mouse CD155 / PVR Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Mouse CD155, Fc Tag, Avi Tag (CD5-M82F7) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 63.5 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse TIGIT, Fc Tag (Cat. No. TIT-M5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse CD155, Fc Tag, Avi Tag (Cat. No. CD5-M82F7) with a linear range of 20-313 ng/mL (QC tested).