Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD155 / PVR Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;CD5,CD155,PVR,FLJ25946,PVS,TAGE4,HVED,NECL5,NECL-5,TAGE-4
Supplier:  ACROBiosystems CD5H82E3200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD155 / PVR Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD155, His Tag, Avi Tag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 38.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIGIT, mouse IgG2a Fc tag, low endotoxin (Cat. No. TIT-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD155, His Tag, Avi Tag (Cat. No. CD5-H82E3) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Biotinylated Human CD155, His Tag, Avi Tag (Cat. No. CD5-H82E3) at 5 μg/mL (100 μL/well) via streptavidin can bind Human TIGIT Protein, Fc Tag (Cat. No. TIT-H5254) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-165-8014