Learn More
Sino Biological Human CD155 / PVR Protein (ECD, Fc & AVI Tag), Biotinylated 20ug
Supplier:  Sino Biological 10109H41HB20
Encompass_Preferred
A DNA sequence encoding the human PVR (P15151-1) (Met1-Asn343) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-6528