Learn More
Enzo Life Sciences CD158a monoclonal antibody (2F9) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7270100
Encompass_Preferred
Clone: 2F9. Isotype: IgG2aκ. Immunogen: Recombinant human kIR2DL1 (aa 23-223) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3268