Learn More
CAPRICO BIOTECHNOLOGIES INC CD16 Antibody, APC Conjugated, Clone 3G8 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4014045
Encompass_Preferred
The Caprico CD16 Antibody (4014045) is an APC-conjugated monoclonal antibody that targets human CD16, also known as Fc gamma receptor III. It is produced in mouse and recognizes an extracellular epitope. This antibody can be used for flow cytometry, immunohistochemistry, and other applications to label and analyze CD16 expression on natural killer cells, monocytes, macrophages, and granulocytes. The immunogen is human granulocytes. This antibody is validated for flow cytometry using human peripheral blood cells.
Catalog No. 50-227-9034