Learn More
CAPRICO BIOTECHNOLOGIES INC CD16 iFluor 700 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4014195
Encompass_Preferred
The CD16 iFluor 700 Antibody from Caprico Biotechnologies is an antibody that targets CD16, also known as Fc gamma receptor III. It is conjugated to iFluor 700, a far-red fluorescent dye with an excitation max of 688nm and emission max of 713nm. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It is available in multiple sizes and formats including 100 tests and 500 tests. According to the product details, this antibody has been tested and validated for flow cytometry applications using human peripheral blood cells.
Catalog No. 50-227-9036