Learn More
CAPRICO BIOTECHNOLOGIES INC CD16 PE Antibody, 4170025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4170025
Encompass_Preferred
The CD16 PE Antibody from Caprico Biotech is a mouse monoclonal antibody that binds to human CD16, also known as Fc gamma receptor III. It is conjugated to R-phycoerythrin (PE) and is intended for use in flow cytometry. This antibody recognizes the CD16a isoform and has been tested for use in analyzing CD16 expression on natural killer cells. According to the product details, this antibody is supplied at a concentration of 0.1 mg/mL in PBS with 0.1% sodium azide and 1% BSA. The immunogen is a purified recombinant protein corresponding to the extracellular domain of human CD16. This antibody can be a useful tool for studying natural killer cell biology and function.
Catalog No. 50-227-9040