Learn More
CAPRICO BIOTECHNOLOGIES INC CD16 PE-Cyanine7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4014085
Encompass_Preferred
The CD16 PE-Cyanine7 antibody is a mouse monoclonal antibody that binds to human CD16, also known as Fc gamma receptor III. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry applications. This antibody recognizes the CD16a and CD16b isoforms. It is recommended for staining human peripheral blood cells. The antibody is supplied at 0.2 mg/mL in PBS with 0.1% sodium azide and 2 mg/mL bovine serum albumin. The clone is 3G8.
Catalog No. 50-227-9038