Learn More
Genscript Corporation CD160 hFc Chimera Mouse
Supplier:  Genscript Corporation Z04296100
Encompass
CD160 (also Natural killer cell receptor BY55) is a 27 30 kDa member of the Ig superfamily In human it is expressed principally on nonmyeloid hematopoietic cells CD160 antigenis a receptor on immune cells capable to deliver stimulatory or inhibitory signals that regulate cell activation and differentiation Exists as a GPI-anchored and as a transmembrane form each likely initiating distinct signaling pathways via phosphoinositol 3-kinase in activated NK cells and via LCK and CD247/CD3 zeta chain in activated T cells
Catalog No. 50-000-57663