Learn More
ACROBiosystems ACROBiosystems Human CD160 Protein, His Tag
Supplier:  ACROBiosystems BY5H52291MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD160, His Tag (BY5-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 159 (Accession # O95971-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human HVEM, Fc Tag (Cat. No. HV4-H82F1) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5μg/well) plate, can bind Human CD160, His Tag (Cat. No. BY5-H5229) with a linear range of 8-63 ng/mL (QC tested).
Catalog No. 50-195-6778