Learn More
Genscript Corporation CD161 hFc Chimera Human
Supplier:  Genscript Corporation Z04161100
Encompass
CD161 (NKRP1) is a lectin-like receptor present on NK cells and rare T-cell subsets We have observed CD161 expression in some cases of T-cell prolymphocytic leukemia (T-PLL) and found it to be useful in follow-up and detection of disease after treatment
Catalog No. 50-000-57781