Learn More
Medchemexpress LLC CD161 Human Biotin 50ug | 50ug
SDP
Catalog No. 5000193377
Encompass_Preferred
CD161 protein plays a crucial role in inhibiting NK cell toxicity by activating specific acid sphingomyelinase/SMPD1 and increasing ceramide levels Activation stimulates AKT1/PKB and RPS6KA1/RSK1 kinases enhancing anti-CD3-induced T cell proliferation CD161 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CD161 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93377 Supplier Medchemexpress LLC Supplier No. HYP7808550UG
CD161 protein plays a crucial role in inhibiting NK cell toxicity by activating specific acid sphingomyelinase/SMPD1 and increasing ceramide levels Activation stimulates AKT1/PKB and RPS6KA1/RSK1 kinases enhancing anti-CD3-induced T cell proliferation CD161 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CD161 protein expressed by HEK293 with C-Avi C-His labeled tag