Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6404MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: PSGL1/1601. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Jurkat cells. Tonsil or Spleen. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length SELPLG (CD162) protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 43-120kDa.Chromosome: 12q24. Gene Symbol: SELPLG. Gene ID: 6404. Ensemble: ENSG00000110876. Uniprot: NP_001193538.
Catalog No. 50-188-5162