Learn More
Enzo Life Sciences CD163 (human) clonal antibody (K20-T) (500µl)
SDP
Supplier:  Enzo Life Sciences ALX810213R500
Encompass_Preferred
Clone: K20-T. Isotype: IgG. Immunogen: Synthetic peptide corresponding to an N-terminal sequence of human CD163. Species reactivity: Human. Applications: IHC (FS), IHC (PS). Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, containing 20mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0080