Learn More
Medchemexpress LLC CD163 Human HEK293 50ug | 50ug
SDP
Catalog No. 5000187790
Encompass_Preferred
The CD163 protein is an acute-phase regulatory receptor that clears hemoglobin/haptoglobin complexes through endocytosis by macrophages protecting tissues from potential oxidative damage In addition to iron uptake CD163 also exhibits calcium- and pH-dependent binding preferences with higher affinity for the hemoglobin/haptoglobin complex of the HP 1F phenotype CD163 Protein Human (HEK293 His-Avi) is the recombinant human-derived CD163 protein expressed by HEK293 with C-His C-Avi labeled tag
Catalog No. 50-001-87790 Supplier Medchemexpress LLC Supplier No. HYP7788450UG
The CD163 protein is an acute-phase regulatory receptor that clears hemoglobin/haptoglobin complexes through endocytosis by macrophages protecting tissues from potential oxidative damage In addition to iron uptake CD163 also exhibits calcium- and pH-dependent binding preferences with higher affinity for the hemoglobin/haptoglobin complex of the HP 1F phenotype CD163 Protein Human (HEK293 His-Avi) is the recombinant human-derived CD163 protein expressed by HEK293 with C-His C-Avi labeled tag