Learn More
ACROBiosystems Recombinant Protein;Human CD163 Protein;HEK293;100UG;CD3,CD163,M130,MM130
Catalog No. 501658961
Encompass_Preferred
Recombinant Protein;Human CD163 Protein;100UG;Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It contains AA Ser 42 - Ser 1045 (Accession # Q86VB7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 110.4 kDa. The protein migrates as 135-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8961 Supplier ACROBiosystems Supplier No. CD3H52E8100UG
Recombinant Protein;Human CD163 Protein;100UG;Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It contains AA Ser 42 - Ser 1045 (Accession # Q86VB7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 110.4 kDa. The protein migrates as 135-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.