Learn More
Cayman Chemical CD163NTerm RabbIt Monclo 100uL
SDP
Catalog No. 502947073
Encompass_Preferred
A Nek2 inhibitor (IC50 56 nM)
Catalog No. 50-294-7073 Supplier Cayman Chemical Supplier No. 32296100 UL
A Nek2 inhibitor (IC50 56 nM)