Learn More
Genscript Corporation CD164 hFc Chimera Mouse
Catalog No. 5000059301
Encompass
CD164 was found to play a role in many malignant diseases CD164 was associated with clinical and pathological features of patients High level of CD164 was related to the distant metastasis and vascular invasion of bladder cancer patients CD164 was associated with the poor clinical outcomes of BC patients Silencing of CD164 could inhibit the progression of tumors in vivo and in vitro which may become an effective target in the treatment of bladder cancer
Catalog No. 50-000-59301 Supplier Genscript Corporation Supplier No. Z04299500
CD164 was found to play a role in many malignant diseases CD164 was associated with clinical and pathological features of patients High level of CD164 was related to the distant metastasis and vascular invasion of bladder cancer patients CD164 was associated with the poor clinical outcomes of BC patients Silencing of CD164 could inhibit the progression of tumors in vivo and in vitro which may become an effective target in the treatment of bladder cancer