Learn More
ABclonal Technology CD166/ALCAM Rabbit mAb
SDP
Catalog No. 5000005276
Encompass_Preferred
This gene encodes activated leukocyte cell adhesion molecule (ALCAM) also known as CD166 (cluster of differentiation 166) which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain This protein binds to T-cell differentiation antigene CD6 and is implicated in the processes of cell adhesion and migration Multiple alternatively spliced transcript variants encoding different isoforms have been found
Catalog No. 50-000-05276 Supplier ABclonal Technology Supplier No. A2344120UL
This gene encodes activated leukocyte cell adhesion molecule (ALCAM) also known as CD166 (cluster of differentiation 166) which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain This protein binds to T-cell differentiation antigene CD6 and is implicated in the processes of cell adhesion and migration Multiple alternatively spliced transcript variants encoding different isoforms have been found