Learn More
Sino Biological Recombinant Human CD16a/FCGR3A/Fc gamma RIIIA Protein (V176, His Tag), HPLC-verified
Supplier:  Sino Biological 10389H08C11
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Gln208) of human FCGR3A (P08637-1, with Natural variant V176)was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-5474