Learn More
Sino Biological Human CD16b / FCGR3B Protein (NA2 allotype, His Tag) 1mg
Supplier:  Sino Biological 11046H08C1
Encompass_Preferred
A DNA sequence encoding the human CD16b (O75015.2) (Met 1-Ser 200) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-211-5436