Learn More
Sino Biological Human CD16B / FCGR3B Protein (His Tag, SH allotype) 100ug
Supplier:  Sino Biological 11046H08H2100
Encompass_Preferred
A DNA sequence encoding the human FCGR3B (NP_000561.3) [(Met 1-Ser 200) (Ala 78 Asp, natural mutation)] was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6305