Learn More
Sino Biological Human CD16b / FCGR3B Protein (NA2 allotype, His & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological 11046H27HB100
Encompass_Preferred
A DNA sequence encoding the human FCGR3B (O75015.2) (Met1-Ser200) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus (AVI-his). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5337