Learn More
CD172a/b Mouse anti-Human, BUV563, Clone: SE5A5, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748435
Description
CD172a/b Monoclonal antibody specifically detects CD172a/b in Human samples. It is validated for Flow Cytometry
Specifications
| CD172a/b | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| SIRP alpha/beta1; SIRPα/SIRPβ1; Signal Regulatory Protein α/β1 | |
| Human SIRP alpha extracellular domain Recombinant Protein | |
| RUO | |
| Human | |
| Mouse BALB/c IgG1 κ |
| Flow Cytometry | |
| SE5A5 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.